Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wt1
Entrez Gene ID
22431
Gene Name
Wilms tumor 1 homolog
Gene Aliases
D630046I19Rik, Wt-1
Chromosome Location
Chr.2: 105126529 - 105173614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144783.2 NP_659032.3 6 - 7 1365 96
GenBank mRNA BG087452.2 - 2 - 3 164 96
DQ537939.1 - 6 - 7 702 96
M55512.1 - 6 - 7 1370 96

View Full Product Details