Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc8
Entrez Gene ID
76260
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
0610012F22Rik, AV001447, BBS8
Chromosome Location
Chr.12: 98920574 - 98983250 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029553.3 NP_083829.1 2 - 3 335 87
NM_198311.1 NP_938053.1 3 - 4 327 87
GenBank mRNA AK002597.1 - 2 - 3 321 87
AK081697.1 - 2 - 3 302 87
BC017523.1 - 3 - 4 327 87

View Full Product Details