Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Efnb3
Entrez Gene ID
13643
Gene Name
ephrin B3
Gene Aliases
EFL-6, ELF-3, Elk-L3, Epl8, LERK-8, NLERK-2
Chromosome Location
Chr.11: 69554092 - 69560595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007911.5 NP_031937.1 3 - 4 938 82
GenBank mRNA AF025288.1 - 3 - 4 556 82
AK048305.1 - 3 - 4 882 82
BC052001.1 - 3 - 4 755 82
BC058617.1 - 3 - 4 871 82

View Full Product Details