Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpv
Entrez Gene ID
73139
Gene Name
centromere protein V
Gene Aliases
3110013H01Rik, AA671303, Prr6
Chromosome Location
Chr.11: 62524944 - 62539502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028448.1 NP_082724.1 1 - 2 411 106
GenBank mRNA AK014046.1 - 1 - 2 411 106
BC147794.1 - 1 - 2 373 106
BC147795.1 - 1 - 2 373 106
BC147835.1 - 1 - 2 373 106
BC147837.1 - 1 - 2 373 106

View Full Product Details