Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigl
Entrez Gene ID
327942
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class L
Gene Aliases
Gm737
Chromosome Location
Chr.11: 62458460 - 62513900 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039536.2 NP_001034625.1 6 - 7 659 107
GenBank mRNA AK128932.1 - 6 - 7 2237 107
AK158149.1 - 6 - 7 660 107

View Full Product Details