Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpv2
Entrez Gene ID
22368
Gene Name
transient receptor potential cation channel, subfamily V, member 2
Gene Aliases
GRC, OTRPC2, VRL-1, Vrl1
Chromosome Location
Chr.11: 62574486 - 62600305 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011706.2 NP_035836.2 9 - 10 1711 71
GenBank mRNA AB021665.1 - 9 - 10 1793 71
AK008689.1 - 7 - 8 1236 71
AK089004.1 - 10 - 11 1726 71
BC005415.1 - 8 - 9 1658 71

View Full Product Details