The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165997.1 | NP_001159469.1 | 2 - 3 | 393 | 140 |
NM_029773.2 | NP_084049.2 | 2 - 3 | 393 | 140 | |
GenBank mRNA | BC111867.1 | - | 2 - 3 | 267 | 140 |
BI648825.1 | - | 2 - 3 | 343 | 140 |