Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spopl
Entrez Gene ID
76857
Gene Name
speckle-type POZ protein-like
Gene Aliases
4921517N04Rik, AU014935, BB233739, E430033K04Rik
Chromosome Location
Chr.2: 23506220 - 23572128 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165997.1 NP_001159469.1 10 - 11 1401 69
NM_001165998.1 NP_001159470.1 9 - 10 1265 69
NM_029773.2 NP_084049.2 9 - 10 1304 69
GenBank mRNA AK014913.1 - 4 - 5 362 69
AK162056.1 - 9 - 10 1119 69
BC111867.1 - 10 - 11 1275 69
CD541905.1 - 4 - 5 398 69

View Full Product Details