Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itih2
Entrez Gene ID
16425
Gene Name
inter-alpha trypsin inhibitor, heavy chain 2
Gene Aliases
AI747202, Intin2, Itih-2
Chromosome Location
Chr.2: 10094591 - 10130683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010582.3 NP_034712.2 18 - 19 2525 92
GenBank mRNA AK133782.1 - 18 - 19 2424 92
BC034341.1 - 18 - 19 2499 92
X70392.1 - 18 - 19 2511 92

View Full Product Details