Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tgfbi
- Entrez Gene ID
-
21810
- Gene Name
- transforming growth factor, beta induced
- Gene Aliases
-
68kDa, AI181842, AI747162, Beta-ig, big-h3
- Chromosome Location
-
Chr.13: 56609603 - 56639339 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC129900.1 |
- |
7 - 8 |
945 |
74 |
View Full Product Details