The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016907.3 | NP_058603.2 | 3 - 4 | 786 | 76 |
GenBank mRNA | AF099018.1 | - | 3 - 4 | 741 | 76 |
AK017342.1 | - | 2 - 3 | 668 | 76 | |
BC005769.1 | - | 2 - 3 | 673 | 76 |