Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
27494
Gene Name
angiomotin
Gene Aliases
CAG-2, D0Kist1, Sii6
Chromosome Location
Chr.X: 145446424 - 145505541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153319.3 NP_695231.3 10 - 11 3282 70
GenBank mRNA AF461135.1 - 10 - 11 2683 70
AK129277.1 - 5 - 6 1522 70
U80888.1 - 1 - 2 854 70

View Full Product Details