Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
27494
Gene Name
angiomotin
Gene Aliases
CAG-2, D0Kist1, Sii6
Chromosome Location
Chr.X: 145446424 - 145505541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290274.1 NP_001277203.1 8 - 9 2173 98
NM_153319.3 NP_695231.3 8 - 9 2173 98
GenBank mRNA AK129277.1 - 3 - 4 413 98
BC057638.1 - 8 - 9 1628 98

View Full Product Details