The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019832.3 | NP_062806.1 | 4 - 5 | 600 | 78 |
GenBank mRNA | AB033130.1 | - | 3 - 4 | 459 | 78 |
AK016588.1 | - | 3 - 4 | 372 | 78 | |
BC052347.1 | - | 4 - 5 | 600 | 78 |