Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
83435
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
FAPP1
Chromosome Location
Chr.2: 76675310 - 76697335 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031256.3 NP_112546.1 6 - 7 936 63
GenBank mRNA AF286163.1 - 6 - 7 725 63
BC012283.1 - 5 - 6 1195 63
BC031110.1 - 7 - 8 744 63

View Full Product Details