The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031256.3 | NP_112546.1 | 7 - 8 | 1052 | 94 |
GenBank mRNA | AF286163.1 | - | 7 - 8 | 841 | 94 |
BC012283.1 | - | 6 - 7 | 1311 | 94 | |
BC031110.1 | - | 8 - 9 | 860 | 94 |