The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164742.1 | NP_001158214.1 | 1 - 2 | 96 | 77 |
NM_026687.2 | NP_080963.2 | 2 - 3 | 236 | 77 | |
GenBank mRNA | AB233454.1 | - | 2 - 3 | 236 | 77 |
AK002574.1 | - | 1 - 2 | 124 | 77 | |
AK165409.1 | - | 1 - 2 | 74 | 77 | |
BC119526.1 | - | 1 - 2 | 64 | 77 |