The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047159.2 | NP_001040624.1 | 5 - 6 | 722 | 74 |
NM_019671.3 | NP_062645.2 | 7 - 8 | 843 | 74 | |
GenBank mRNA | AF094520.1 | - | 7 - 8 | 739 | 74 |
AJ010045.1 | - | 5 - 6 | 685 | 74 | |
AK082128.1 | - | 5 - 6 | 719 | 74 | |
AK088632.1 | - | 5 - 6 | 686 | 74 | |
AK139921.1 | - | 5 - 6 | 686 | 74 | |
BC004699.1 | - | 5 - 6 | 696 | 74 |