The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033167.3 | NP_001028339.1 | 1 - 2 | 1025 | 133 |
GenBank mRNA | AK147397.1 | - | 1 - 2 | 1026 | 133 |