Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a23
Entrez Gene ID
73102
Gene Name
solute carrier family 22, member 23
Gene Aliases
3110004L20Rik
Chromosome Location
Chr.13: 34179158 - 34345487 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC039219.1 - 5 - 6 519 68
BC041671.1 - 5 - 6 519 68
BC053705.1 - 6 - 7 738 68
BC094372.1 - - 714 68

View Full Product Details