Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shc3
Entrez Gene ID
20418
Gene Name
src homology 2 domain-containing transforming protein C3
Gene Aliases
N-Shc, Rai, ShcC
Chromosome Location
Chr.13: 51431041 - 51567084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009167.3 NP_033193.2 3 - 4 254 66
GenBank mRNA BC103611.1 - 3 - 4 254 66
BC103612.1 - 3 - 4 254 66
BC105644.1 - 3 - 4 254 66
BC105645.1 - 3 - 4 254 66
U46854.1 - 3 - 4 254 66

View Full Product Details