The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018859.2 | NP_061347.2 | 8 - 9 | 817 | 90 |
GenBank mRNA | AK002507.1 | - | 8 - 9 | 792 | 90 |
AK028605.1 | - | - | 643 | 90 | |
BC012692.1 | - | 8 - 9 | 833 | 90 | |
U68535.1 | - | 8 - 9 | 803 | 90 |