Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akr1c21
Entrez Gene ID
77337
Gene Name
aldo-keto reductase family 1, member C21
Gene Aliases
9430025F20Rik, AI315367
Chromosome Location
Chr.13: 4574075 - 4586543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029901.2 NP_084177.2 4 - 5 518 126
GenBank mRNA AB178898.1 - 4 - 5 444 126
BC013531.1 - 4 - 5 494 126
BC061057.1 - 4 - 5 514 126
BC091761.1 - 4 - 5 494 126

View Full Product Details