The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152824.1 | NP_690037.1 | 11 - 12 | 1240 | 95 |
GenBank mRNA | AF083384.1 | - | 10 - 11 | 1110 | 95 |
AK012296.1 | - | 11 - 12 | 1186 | 95 | |
BC034896.1 | - | 11 - 12 | 1240 | 95 |