Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clptm1l
Entrez Gene ID
218335
Gene Name
CLPTM1-like
Gene Aliases
C130052I12Rik, Crr9
Chromosome Location
Chr.13: 73604002 - 73620639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146047.2 NP_666159.2 15 - 16 1656 125
GenBank mRNA AK032295.1 - 15 - 16 1620 125
AK048357.1 - 15 - 16 1553 125
AK156203.1 - 15 - 16 1547 125
AK165939.1 - 15 - 16 1517 125
AK181076.1 - 7 - 8 378 125
AK187892.1 - 7 - 8 362 125
BC026562.1 - 15 - 16 1357 125

View Full Product Details