Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc6a3
Entrez Gene ID
13162
Gene Name
solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene Aliases
DAT, Dat1
Chromosome Location
Chr.13: 73535528 - 73578672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010020.3 NP_034150.1 6 - 7 1080 74
GenBank mRNA AF109072.1 - 6 - 7 1003 74
AF109391.1 - 5 - 6 928 74
AJ238309.1 - 5 - 6 936 74
AK032629.1 - 6 - 7 1038 74
AK136885.1 - 6 - 7 1025 74
BC054119.1 - 6 - 7 1080 74

View Full Product Details