Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insl3
Entrez Gene ID
16336
Gene Name
insulin-like 3
Gene Aliases
Rlf, Rlnl
Chromosome Location
Chr.8: 71689252 - 71690577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013564.7 NP_038592.3 1 - 2 182 149
GenBank mRNA AK132857.1 - 1 - 2 183 149
BC049540.1 - 1 - 2 182 149
S82815.1 - 1 - 2 181 149

View Full Product Details