The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021506.2 | NP_067481.2 | 2 - 3 | 662 | 118 |
GenBank mRNA | AF030131.1 | - | 2 - 3 | 561 | 118 |
AK088003.1 | - | 2 - 3 | 602 | 118 | |
AK148137.1 | - | 2 - 3 | 590 | 118 | |
AK173185.1 | - | 2 - 3 | 1025 | 118 | |
BC060113.1 | - | 2 - 3 | 624 | 118 | |
BC060696.1 | - | 2 - 3 | 624 | 118 |