Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drp2
Entrez Gene ID
13497
Gene Name
dystrophin related protein 2
Gene Aliases
AW495265, DRP-2
Chromosome Location
Chr.X: 134404628 - 134456574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010078.3 NP_034208.2 4 - 5 667 68
GenBank mRNA AK034383.1 - 4 - 5 610 68
AK081426.1 - 4 - 5 611 68
AK158102.1 - 3 - 4 469 68
BC060605.1 - 3 - 4 438 68
BC125345.1 - 4 - 5 527 68
BC125347.1 - 4 - 5 527 68

View Full Product Details