Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mctp1
Entrez Gene ID
78771
Gene Name
multiple C2 domains, transmembrane 1
Gene Aliases
2810465F10Rik
Chromosome Location
Chr.13: 76384961 - 77033284 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030174.2 NP_084450.2 10 - 11 1363 96
GenBank mRNA AK047562.1 - 10 - 11 1364 96
AK054478.1 - 6 - 7 736 96

View Full Product Details