Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mctp1
Entrez Gene ID
78771
Gene Name
multiple C2 domains, transmembrane 1
Gene Aliases
2810465F10Rik
Chromosome Location
Chr.13: 76384961 - 77033284 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030174.2 NP_084450.2 15 - 16 1867 127
GenBank mRNA AK013379.1 - 2 - 3 473 127
AK047562.1 - 15 - 16 1868 127

View Full Product Details