Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mctp1
Entrez Gene ID
78771
Gene Name
multiple C2 domains, transmembrane 1
Gene Aliases
2810465F10Rik
Chromosome Location
Chr.13: 76384961 - 77033284 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030174.2 NP_084450.2 2 - 3 407 119
GenBank mRNA AK047562.1 - 2 - 3 408 119
AK144765.1 - 2 - 3 206 119
AK154322.1 - 2 - 3 261 119

View Full Product Details