Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks6
Entrez Gene ID
75691
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
2210417J20Rik, Ankrd14, Gm635, SamCystin, Samd6, b2b1801.1Clo
Chromosome Location
Chr.4: 47015365 - 47057335 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024136.1 NP_001019307.1 11 - 12 2144 82
GenBank mRNA BC060663.1 - 12 - 13 2348 82
BC072562.1 - 5 - 6 735 82

View Full Product Details