Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lars2
Entrez Gene ID
102436
Gene Name
leucyl-tRNA synthetase, mitochondrial
Gene Aliases
AI035546, Kiaa0028, LEURS
Chromosome Location
Chr.9: 123366940 - 123462666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153168.2 NP_694808.1 2 - 3 413 69
GenBank mRNA AJ428066.1 - 2 - 3 413 69
AK032695.1 - 2 - 3 418 69
AK037012.1 - 2 - 3 420 69
AK045640.1 - 2 - 3 372 69
AK053667.1 - 2 - 3 407 69
BC053916.1 - 2 - 3 399 69
BC080303.1 - 2 - 3 300 69
BC111892.1 - 2 - 3 384 69
BC112412.1 - 2 - 3 382 69
BC138206.1 - 2 - 3 387 69
BC145982.1 - 2 - 3 387 69

View Full Product Details