Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Timp1
Entrez Gene ID
21857
Gene Name
tissue inhibitor of metalloproteinase 1
Gene Aliases
Clgi, EPA, TIMP-1, TPA-S1, Timp
Chromosome Location
Chr.X: 20870166 - 20874737 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044384.1 NP_001037849.1 4 - 5 501 72
NM_001294280.2 NP_001281209.1 3 - 4 421 72
NM_011593.2 NP_035723.2 4 - 5 498 72
GenBank mRNA AA184230.1 - 2 - 3 267 72
AY622853.1 - 1 - 2 35 72
BC008107.1 - 4 - 5 403 72
BC034260.1 - 4 - 5 403 72
BC051260.1 - 4 - 5 383 72
BU756519.1 - 2 - 3 72
M17243.1 - 4 - 5 364 72
X04684.1 - 4 - 5 440 72

View Full Product Details