The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044384.1 | NP_001037849.1 | 5 - 6 | 618 | 100 |
NM_001294280.2 | NP_001281209.1 | 4 - 5 | 538 | 100 | |
NM_011593.2 | NP_035723.2 | 5 - 6 | 615 | 100 | |
GenBank mRNA | AA184230.1 | - | 3 - 4 | 384 | 100 |
AY622853.1 | - | 2 - 3 | 152 | 100 | |
BC008107.1 | - | 5 - 6 | 520 | 100 | |
BC034260.1 | - | 5 - 6 | 520 | 100 | |
BC051260.1 | - | 5 - 6 | 500 | 100 | |
BU756519.1 | - | 1 - 2 | 100 | ||
M17243.1 | - | 5 - 6 | 481 | 100 | |
X04684.1 | - | 5 - 6 | 557 | 100 |