The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020049.4 | NP_064433.3 | 1 - 2 | 300 | 113 |
GenBank mRNA | AB033285.1 | - | 1 - 2 | 58 | 113 |
AF161714.1 | - | 1 - 2 | 300 | 113 | |
AF320226.1 | - | 1 - 2 | 244 | 113 | |
AK018553.1 | - | 1 - 2 | 159 | 113 | |
BC129908.1 | - | 1 - 2 | 91 | 113 | |
BC129909.1 | - | 1 - 2 | 91 | 113 |