Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stt3b
Entrez Gene ID
68292
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog B (S. cerevisiae)
Gene Aliases
1300006C19Rik, Simp
Chromosome Location
Chr.9: 115242581 - 115310421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024222.2 NP_077184.2 15 - 16 2761 145
GenBank mRNA AK012153.1 - 15 - 16 2431 145
AK018758.2 - 15 - 16 2471 145
AK145674.1 - 15 - 16 2402 145
AK148065.1 - 15 - 16 2460 145
AK152899.1 - 15 - 16 2809 145
AK154979.1 - 15 - 16 2450 145
AK170079.1 - 15 - 16 2473 145
AX799084.1 - 15 - 16 2472 145
BC003206.1 - 10 - 11 1473 145
BC013054.1 - 5 - 6 879 145
BC052433.1 - 15 - 16 2761 145

View Full Product Details