Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trmt10b
Entrez Gene ID
69934
Gene Name
tRNA methyltransferase 10B
Gene Aliases
2610042J10Rik, Rg9mtd3, Rnmtd3
Chromosome Location
Chr.4: 45297071 - 45316131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027266.4 NP_081542.2 8 - 9 907 75
GenBank mRNA AK011746.1 - 8 - 9 920 75
AK030465.1 - 8 - 9 908 75
BC019428.1 - 7 - 8 884 75

View Full Product Details