The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175114.3 | NP_780323.2 | 15 - 16 | 2205 | 79 |
GenBank mRNA | AK029123.1 | - | 17 - 18 | 2674 | 79 |
BC058971.1 | - | 15 - 16 | 2147 | 79 | |
BC082807.1 | - | 2 - 3 | 171 | 79 |