The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037761.2 | NP_001032850.1 | 3 - 4 | 342 | 94 |
NM_001271405.1 | NP_001258334.1 | 3 - 4 | 321 | 94 | |
NM_001271406.1 | NP_001258335.1 | 3 - 4 | 307 | 94 | |
NM_009798.4 | NP_033928.1 | 3 - 4 | 342 | 94 | |
GenBank mRNA | AK153367.1 | - | 3 - 4 | 273 | 94 |
AK153394.1 | - | 3 - 4 | 273 | 94 | |
AK156778.1 | - | 3 - 4 | 311 | 94 | |
AK159390.1 | - | 3 - 4 | 316 | 94 | |
AK160082.1 | - | 3 - 4 | 317 | 94 | |
AK162757.1 | - | 3 - 4 | 309 | 94 | |
AK168738.1 | - | 3 - 4 | 270 | 94 | |
BC002053.1 | - | 3 - 4 | 284 | 94 | |
FJ692320.1 | - | 3 - 4 | 307 | 94 | |
U10406.1 | - | 3 - 4 | 262 | 94 | |
U10407.1 | - | 3 - 4 | 233 | 94 |