Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dbn1
Entrez Gene ID
56320
Gene Name
drebrin 1
Gene Aliases
-
Chromosome Location
Chr.13: 55473428 - 55488163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177371.1 NP_001170842.1 3 - 4 381 60
NM_001177372.1 NP_001170843.1 3 - 4 381 60
NM_019813.4 NP_062787.2 3 - 4 381 60
GenBank mRNA AK132060.1 - 3 - 4 367 60
AK162697.1 - 3 - 4 368 60
BC006714.1 - 4 - 5 329 60

View Full Product Details