Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hk3
Entrez Gene ID
212032
Gene Name
hexokinase 3
Gene Aliases
HK III, HK-III
Chromosome Location
Chr.13: 55005985 - 55021472 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033245.4 NP_001028417.1 11 - 12 1857 69
NM_001206390.1 NP_001193319.1 11 - 12 1667 69
NM_001206391.1 NP_001193320.1 11 - 12 1532 69
NM_001206392.1 NP_001193321.1 10 - 11 1737 69
GenBank mRNA AK140444.1 - 11 - 12 1721 69
AK149651.1 - 11 - 12 1707 69
AK149992.1 - 11 - 12 1669 69
AK152839.1 - 5 - 6 961 69
AK154603.1 - 11 - 12 1829 69
AK155701.1 - 11 - 12 1714 69
AK159857.1 - 10 - 11 1583 69
AK159906.1 - 11 - 12 1767 69
AK160018.1 - 11 - 12 1767 69
AK160063.1 - 11 - 12 1767 69
AK162637.1 - 11 - 12 1800 69
AK163658.1 - 11 - 12 1721 69
AK170813.1 - 10 - 11 1587 69
AK171214.1 - 11 - 12 1795 69
AK171248.1 - 11 - 12 1707 69
AK171580.1 - 11 - 12 1807 69

View Full Product Details