Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hk3
Entrez Gene ID
212032
Gene Name
hexokinase 3
Gene Aliases
HK III, HK-III
Chromosome Location
Chr.13: 55005985 - 55021472 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033245.4 NP_001028417.1 13 - 14 2120 69
NM_001206390.1 NP_001193319.1 13 - 14 1930 69
NM_001206391.1 NP_001193320.1 13 - 14 1795 69
NM_001206392.1 NP_001193321.1 12 - 13 2000 69
GenBank mRNA AK140444.1 - 13 - 14 1984 69
AK149651.1 - 13 - 14 1970 69
AK149992.1 - 13 - 14 1932 69
AK152839.1 - 7 - 8 1224 69
AK154603.1 - 13 - 14 2092 69
AK155701.1 - 13 - 14 1978 69
AK159857.1 - 12 - 13 1846 69
AK159906.1 - 13 - 14 2030 69
AK160018.1 - 13 - 14 2030 69
AK160063.1 - 13 - 14 2030 69
AK162637.1 - 13 - 14 2063 69
AK163658.1 - 13 - 14 1984 69
AK170813.1 - 12 - 13 1850 69
AK171214.1 - 13 - 14 2058 69
AK171248.1 - 13 - 14 1970 69
AK171580.1 - 13 - 14 2070 69
BC117860.1 - 13 - 14 2015 69
BC117861.1 - 13 - 14 1880 69

View Full Product Details