Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmrtc1c2
Entrez Gene ID
668357
Gene Name
DMRT-like family C1c2
Gene Aliases
-
Chromosome Location
Chr.X: 102847636 - 102857207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142690.1 NP_001136162.1 2 - 3 385 64

Gene Symbol
Dmrtc1c1
Entrez Gene ID
71083
Gene Name
DMRT-like family C1c1
Gene Aliases
4933424F23Rik, Dmrt8.3, Dmrtc1c
Chromosome Location
Chr.X: 102797880 - 102809945 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142691.1 NP_001136163.1 2 - 3 385 64
GenBank mRNA AK016891.1 - 2 - 3 386 64

View Full Product Details