The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170978.1 | NP_001164449.1 | 10 - 11 | 853 | 73 |
NM_172961.3 | NP_766549.2 | 10 - 11 | 853 | 73 | |
GenBank mRNA | AK036128.1 | - | 10 - 11 | 798 | 73 |
AK132282.1 | - | 10 - 11 | 802 | 73 | |
AK165348.1 | - | 10 - 11 | 803 | 73 | |
BC058079.1 | - | 10 - 11 | 793 | 73 | |
BC058521.1 | - | 10 - 11 | 793 | 73 |