The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009541.2 | NP_033567.2 | 15 - 16 | 2322 | 66 |
GenBank mRNA | AK051971.1 | - | 14 - 15 | 2393 | 66 |
AK134251.1 | - | 15 - 16 | 2282 | 66 | |
BC060055.1 | - | 12 - 13 | 2592 | 66 | |
BC145312.1 | - | 14 - 15 | 2195 | 66 | |
BC145821.1 | - | 14 - 15 | 2195 | 66 | |
U14556.1 | - | 15 - 16 | 2320 | 66 | |
U22396.1 | - | 15 - 16 | 2310 | 66 |