The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122667.2 | NP_001116139.1 | 16 - 17 | 2912 | 79 |
NM_153588.3 | NP_705816.2 | 14 - 15 | 2926 | 79 | |
GenBank mRNA | AF532598.1 | - | 14 - 15 | 2708 | 79 |
AK220377.1 | - | 14 - 15 | 2708 | 79 | |
BC131644.1 | - | 14 - 15 | 2708 | 79 | |
BC131645.1 | - | 14 - 15 | 2706 | 79 | |
CK639342.1 | - | 2 - 3 | 96 | 79 |