The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182839.2 | NP_878259.1 | 1 - 2 | 92 | 97 |
GenBank mRNA | BY124320.1 | - | 1 - 2 | 92 | 97 |