The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101588.1 | NP_001095058.1 | 1 - 2 | 199 | 75 |
GenBank mRNA | BC158089.1 | - | 1 - 2 | 276 | 75 |